This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04493070_g1
See other ATP5H GE Assays ›
Gene Symbol:
Atp5h
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003613598: 471553 - 476371 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04493070_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5h
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003495664.2 XP_003495712.1 5 - 6 408 109
GenBank mRNA JI866436.1 - 5 - 6 416 109
JP044311.1 - 5 - 6 416 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Transporter 3' Most Detect Genomic DNA Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0