This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04493938_m1
See other EXTL2 GE Assays ›
Gene Symbol:
Extl2
Gene Name:
exostosin-like glycosyltransferase 2
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003615591: 81552 - 104865 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04493938_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Extl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007653208.1 XP_007651398.1 3 - 4 722 74
XM_007653209.1 XP_007651399.1 4 - 5 883 74
XM_007653210.1 XP_007651400.1 4 - 5 905 74
XM_007653211.1 XP_007651401.1 3 - 4 701 74
XM_007653212.1 XP_007651402.1 4 - 5 888 74
XM_007653213.1 XP_007651403.1 5 - 6 1006 74
XM_007653214.1 XP_007651404.1 4 - 5 840 74
XM_007653215.1 XP_007651405.1 3 - 4 811 74
XM_007653216.1 XP_007651406.1 2 - 3 645 74
GenBank mRNA JP064350.1 - 1 - 2 435 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0