This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04507262_m1
See other SEC22C GE Assays ›
Gene Symbol:
Sec22c
Gene Name:
SEC22 vesicle trafficking protein homolog C (S. cerevisiae)
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003613803: 1176560 - 1202434 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04507262_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec22c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003500691.2 XP_003500739.1 2 - 3 595 88
XM_007641435.1 XP_007639625.1 2 - 3 595 88
XM_007641436.1 XP_007639626.1 2 - 3 553 88
XM_007641437.1 XP_007639627.1 2 - 3 1234 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Inventoried Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0