This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04513310_g1
See other MUTYH GE Assays ›
Gene Symbol:
Mutyh
Gene Name:
mutY homolog
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003613647: 2368844 - 2378831 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04513310_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mutyh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007611343.1 XP_007609533.1 10 - 11 1016 60
XM_007611346.1 XP_007609536.1 11 - 12 1108 60
XM_007611354.1 XP_007609544.1 11 - 12 1013 60
XM_007611361.1 XP_007609551.1 10 - 11 964 60
XM_007611370.1 XP_007609560.1 9 - 10 878 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

DNA glycosylase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-7zhdl:80/100.66.77.60:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0