This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04518395_m1
See other NXPE4 GE Assays ›
Gene Symbol:
Nxpe4
Gene Name:
neurexophilin and PC-esterase domain family, member 4
Gene Aliases:
I79_017457
Chromosome Location:
Chr.Un|NW_003614687: 473141 - 516463 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04518395_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nxpe4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003509321.2 XP_003509369.1 7 - 8 1372 66
XM_007649899.1 XP_007648089.1 9 - 10 1699 66
XM_007649900.1 XP_007648090.1 9 - 10 1822 66
XM_007649901.1 XP_007648091.1 10 - 11 1926 66
XM_007649902.1 XP_007648092.1 7 - 8 1563 66
XM_007649903.1 XP_007648093.1 9 - 10 1691 66
XM_007649904.1 XP_007648094.1 7 - 8 1440 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline