This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04518601_g1
See other DFFB GE Assays ›
Gene Symbol:
Dffb
Gene Name:
DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase)
Gene Aliases:
I79_024699
Chromosome Location:
Chr.Un|NW_003617576: 17897 - 30770 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04518601_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dffb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003515229.2 XP_003515277.1 3 - 4 587 66
XM_007655221.1 XP_007653411.1 3 - 4 591 66
XM_007655222.1 XP_007653412.1 3 - 4 590 66
XM_007655223.1 XP_007653413.1 3 - 4 594 66
XM_007655224.1 XP_007653414.1 3 - 4 593 66
XM_007655225.1 XP_007653415.1 3 - 4 593 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Apoptosis Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0