This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04518610_g1
See other FCGRT GE Assays ›
Gene Symbol:
Fcgrt
Gene Name:
Fc fragment of IgG, receptor, transporter, alpha
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003617747: 19705 - 28718 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04518610_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fcgrt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003515304.2 XP_003515352.1 6 - 7 1171 70
XM_007607530.1 XP_007605720.1 6 - 7 1362 70
XM_007607531.1 XP_007605721.1 6 - 7 1284 70
XM_007607533.1 XP_007605723.1 5 - 6 1092 70
GenBank mRNA JI889354.1 - 2 - 3 291 70
JP045000.1 - 2 - 3 291 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

major histocompatibility complex protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bwr8z:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0