This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04523577_g1
See other ATP5B GE Assays ›
Gene Symbol:
Atp5b
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003614217: 171492 - 178825 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04523577_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003506176.2 XP_003506224.1 8 - 9 1324 121
GenBank mRNA JI874525.1 - 7 - 8 1089 121
JP060300.1 - 7 - 8 1089 121

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Ion Channel Transporter Detect Genomic DNA Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0