This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04524892_m1
See other NFE2L2 GE Assays ›
Gene Symbol:
Nfe2l2
Gene Name:
nuclear factor, erythroid 2-like 2
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003613688: 418031 - 444597 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04524892_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nfe2l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003498316.2 XP_003498364.2 4 - 5 567 65
XM_007626148.1 XP_007624338.1 4 - 5 741 65
XM_007626157.1 XP_007624347.1 4 - 5 777 65
XM_007626168.1 XP_007624358.1 4 - 5 745 65
XM_007626176.1 XP_007624366.1 4 - 5 678 65
XM_007626185.1 XP_007624375.1 4 - 5 745 65
GenBank mRNA JI867791.1 - 1 - 2 185 65
JP056420.1 - 2 - 3 284 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline