This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04531999_m1
See other DUSP15 GE Assays ›
Gene Symbol:
Dusp15
Gene Name:
dual specificity phosphatase 15
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003613890: 1099060 - 1108390 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04531999_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dusp15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003502323.2 XP_003502371.1 3 - 4 236 89
XM_007642920.1 XP_007641110.1 3 - 4 236 89
XM_007642921.1 XP_007641111.1 3 - 4 334 89
XM_007642922.1 XP_007641112.1 3 - 4 236 89
XM_007642923.1 XP_007641113.1 2 - 3 202 89
GenBank mRNA JI875547.1 - 3 - 4 203 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_007642923.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Protein Phosphatase 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0