This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04536875_m1
See other TMPRSS11A GE Assays ›
Gene Symbol:
Tmprss11a
Gene Name:
transmembrane protease, serine 11A
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003614886: 50600 - 83161 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04536875_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss11a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007651102.1 XP_007649292.1 5 - 6 504 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Protease Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-8rs9g:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline