This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04545708_m1
See other TAP1 GE Assays ›
Gene Symbol:
Tap1
Gene Name:
transporter 1, ATP-binding cassette, sub-family B (MDR/TAP)
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003614196: 865633 - 876434 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04545708_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007646392.1 XP_007644582.1 2 - 3 1012 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Transporter Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline