This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04562499_m1
See other CDKL4 GE Assays ›
Gene Symbol:
Cdkl4
Gene Name:
cyclin-dependent kinase-like 4
Gene Aliases:
I79_016413
Chromosome Location:
Chr.Un|NW_003614547: 194293 - 227524 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04562499_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdkl4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003508459.2 XP_003508507.1 8 - 9 965 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Protein Kinase 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline