Assay Details
| Gene Symbol: Noc4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642386.1 | XP_007640576.1 | 11 - 12 | 999 | 59 | |
| GenBank mRNA | JI871101.1 | - | 11 - 12 | 1117 | 59 | |
More Information
Set Membership: |
|

Noc4l
Genomic Map