This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04571433_m1
See other PCMTD1 GE Assays ›
Gene Symbol:
Pcmtd1
Gene Name:
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003614376: 674944 - 726907 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04571433_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcmtd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003507389.2 XP_003507437.1 4 - 5 717 101
XM_007647985.1 XP_007646175.1 4 - 5 758 101
XM_007647986.1 XP_007646176.1 4 - 5 948 101
XM_007647987.1 XP_007646177.1 2 - 3 1104 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-s62d2:80/100.66.75.204:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0