This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04574856_m1
See other TMEM9 GE Assays ›
Gene Symbol:
Tmem9
Gene Name:
transmembrane protein 9
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003613706: 1847126 - 1865221 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04574856_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003498868.2 XP_003498916.1 5 - 6 488 72
XM_007634537.1 XP_007632727.1 5 - 6 497 72
XM_007634543.1 XP_007632733.1 5 - 6 576 72
XM_007634553.1 XP_007632743.1 5 - 6 565 72
GenBank mRNA JI873531.1 - 5 - 6 497 72
JP050649.1 - 5 - 6 496 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline