This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04591019_m1
See other PPP4C GE Assays ›
Gene Symbol:
Ppp4c
Gene Name:
protein phosphatase 4, catalytic subunit
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003614830: 98201 - 103797 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04591019_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp4c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003510102.2 XP_003510150.1 8 - 9 960 62
GenBank mRNA JI880023.1 - 8 - 9 933 62
JP059397.1 - 8 - 9 933 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Protein Phosphatase 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0