Assay Details
| Gene Symbol: Ogfod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496913.2 | XP_003496961.1 | 4 - 5 | 452 | 60 | |
| XM_007654296.1 | XP_007652486.1 | 4 - 5 | 452 | 60 | ||
| XM_007654305.1 | XP_007652495.1 | 4 - 5 | 357 | 60 | ||
| GenBank mRNA | JI891386.1 | - | 4 - 5 | 401 | 60 | |
More Information
Set Membership: |
|

Ogfod3
Genomic Map