Assay Details
| Gene Symbol: LOC100751727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502309.1 | XP_003502357.1 | 3 - 4 | 356 | 73 | |
| XM_003502310.1 | XP_003502358.1 | 3 - 4 | 356 | 73 | ||
| XM_003502311.1 | XP_003502359.1 | 3 - 4 | 356 | 73 | ||
| XM_007642913.1 | XP_007641103.1 | 4 - 5 | 386 | 73 | ||
More Information
Set Membership: |
|

LOC100751727
Genomic Map