This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04627101_m1
See other CALHM3 GE Assays ›
Gene Symbol:
Calhm3
Gene Name:
calcium homeostasis modulator 3
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003615368: 90463 - 101037 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04627101_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Calhm3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003512142.2 XP_003512190.2 4 - 5 835 78
XM_007652627.1 XP_007650817.1 4 - 5 1167 78
XM_007652628.1 XP_007650818.1 4 - 5 937 78
XM_007652629.1 XP_007650819.1 4 - 5 1276 78
XM_007652630.1 XP_007650820.1 4 - 5 2026 78
XM_007652631.1 XP_007650821.1 3 - 4 1828 78
XM_007652632.1 XP_007650822.1 3 - 4 1598 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mmql6:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0