This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04632496_m1
See other SH3BGR GE Assays ›
Gene Symbol:
Sh3bgr
Gene Name:
SH3 domain binding glutamate-rich protein
Gene Aliases:
I79_022168
Chromosome Location:
Chr.Un|NW_003615800: 52973 - 88286 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04632496_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sh3bgr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003513237.2 XP_003513285.2 3 - 4 513 59
XM_007653535.1 XP_007651725.1 4 - 5 586 59
XM_007653536.1 XP_007651726.1 4 - 5 586 59
XM_007653537.1 XP_007651727.1 3 - 4 535 59
XM_007653538.1 XP_007651728.1 5 - 6 439 59
XM_007653539.1 XP_007651729.1 4 - 5 465 59
XM_007653540.1 XP_007651730.1 4 - 5 522 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-6zkmz:80/100.66.79.198:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline