This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04633589_m1
See other FH GE Assays ›
Gene Symbol:
Fh
Gene Name:
fumarate hydratase
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003613756: 1123557 - 1146615 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04633589_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003499844.2 XP_003499892.1 4 - 5 579 58
XM_007640597.1 XP_007638787.1 5 - 6 735 58
GenBank mRNA JI889546.1 - 3 - 4 422 58
JP055734.1 - 3 - 4 422 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Inventoried Probe spans exons

Panther Classification:

Molecular Function -

lyase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0