This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04634459_m1
See other ZCCHC17 GE Assays ›
Gene Symbol:
Zcchc17
Gene Name:
zinc finger, CCHC domain containing 17
Gene Aliases:
I79_004155
Chromosome Location:
Chr.Un|NW_003613689: 394003 - 441475 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04634459_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zcchc17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007626539.1 XP_007624729.1 6 - 7 554 69
XM_007626546.1 XP_007624736.1 6 - 7 504 69
XM_007626553.1 XP_007624743.1 6 - 7 610 69
XM_007626562.1 XP_007624752.1 5 - 6 429 69
XM_007626571.1 XP_007624761.1 5 - 6 494 69
XM_007626577.1 XP_007624767.1 4 - 5 355 69
GenBank mRNA JI880703.1 - 1 - 2 102 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0