Assay Details
| Gene Symbol: Dbi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499446.2 | XP_003499494.1 | 3 - 4 | 396 | 129 | |
| GenBank mRNA | JI880680.1 | - | 3 - 4 | 354 | 129 | |
More Information
Set Membership: |
|

Dbi
Genomic Map