This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch00610967_m1
See other YIF1A GE Assays ›
Gene Symbol:
YIF1A
Gene Name:
Yip1 interacting factor homolog A, membrane trafficking protein
Gene Aliases:
-
Chromosome Location:
Chr.29: 44741692 - 44745781 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch00610967_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YIF1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005699918.3 XP_005699975.1 1 - 2 274 90
XM_013976041.1 XP_013831495.1 1 - 2 264 90
GenBank mRNA JO413022.2 - 7 - 8 904 90
JO440047.2 - 1 - 2 198 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0