This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch02663798_gH
See other RBBP7 GE Assays ›
Gene Symbol:
RBBP7
Gene Name:
RB binding protein 7, chromatin remodeling factor
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_017190135: 8734 - 30178 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch02663798_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBBP7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018045072.1 XP_017900561.1 2 - 3 503 67
XM_018045073.1 XP_017900562.1 2 - 3 479 67
GenBank mRNA JO581151.1 - 10 - 11 1550 67
KA326291.1 - 10 - 11 1476 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-782pg:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline