This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03211197_g1
See other TMEM183A GE Assays ›
Gene Symbol:
TMEM183A
Gene Name:
transmembrane protein 183A
Gene Aliases:
-
Chromosome Location:
Chr.16: 107992 - 123456 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03211197_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM183A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005690361.2 XP_005690418.1 2 - 3 344 64
XM_005690362.3 XP_005690419.1 2 - 3 353 64
XM_005690363.3 XP_005690420.1 2 - 3 344 64
XM_005690364.3 XP_005690421.1 2 - 3 344 64
XM_018059990.1 XP_017915479.1 2 - 3 344 64
XM_018059991.1 XP_017915480.1 2 - 3 353 64
XM_018059992.1 XP_017915481.1 2 - 3 344 64
XM_018059993.1 XP_017915482.1 2 - 3 353 64
GenBank mRNA JO577711.1 - 6 - 7 1516 64
KA317566.1 - 2 - 3 336 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0