This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03211283_m1
See other MOB3C GE Assays ›
Gene Symbol:
MOB3C
Gene Name:
MOB kinase activator 3C
Gene Aliases:
-
Chromosome Location:
Chr.3: 21243893 - 21252656 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03211283_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOB3C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005678502.3 XP_005678559.1 1 - 2 1735 68
XM_018043797.1 XP_017899286.1 2 - 3 644 68
GenBank mRNA JO598610.2 - 2 - 3 2169 68
JT687384.1 - 2 - 3 544 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

kinase activator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0