This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03225185_g1
See other UCP2 GE Assays ›
Gene Symbol:
UCP2
Gene Name:
uncoupling protein 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 29614180 - 29620957 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03225185_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001314147.1 NP_001301076.1 5 - 6 844 57
XM_018058858.1 XP_017914347.1 7 - 8 1365 57
GenBank mRNA JN986832.1 - 5 - 6 844 57
JO365672.2 - 4 - 5 668 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-9bd97d8-8bg8g:80/100.66.75.22:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline