This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03228749_g1
See other PPP1R2 GE Assays ›
Gene Symbol:
PPP1R2
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 71839911 - 71861038 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03228749_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005675094.3 XP_005675151.1 5 - 6 902 72
XM_018046472.1 XP_017901961.1 5 - 6 905 72
GenBank mRNA JO323288.2 - 1 - 2 408 72
JO323289.2 - 1 - 2 408 72
JO358084.2 - 5 - 6 809 72
JO358085.2 - 5 - 6 810 72
JO373202.2 - 5 - 6 802 72
JO410357.2 - 1 - 2 397 72
JO428626.2 - 1 - 2 1468 72
JO428638.2 - 1 - 2 1466 72
JO502043.2 - 5 - 6 865 72
JO539014.2 - 5 - 6 857 72
JO539016.2 - 5 - 6 858 72
JO562388.2 - 1 - 2 347 72
JO612719.2 - 1 - 2 395 72
JO645102.2 - 5 - 6 899 72
KA346344.1 - 5 - 6 889 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase inhibitor phosphatase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0