This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03229398_m1
See other GTF2A2 GE Assays ›
Gene Symbol:
GTF2A2
Gene Name:
general transcription factor IIA subunit 2
Gene Aliases:
-
Chromosome Location:
Chr.10: 52515367 - 52548027 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03229398_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005685699.3 XP_005685756.1 3 - 4 442 75
XM_005685700.3 XP_005685757.1 3 - 4 378 75
GenBank mRNA JO613354.2 - 3 - 4 359 75
KA342278.1 - 2 - 3 362 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline