This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03241742_m1
See other NAA60 GE Assays ›
Gene Symbol:
NAA60
Gene Name:
N(alpha)-acetyltransferase 60, NatF catalytic subunit
Gene Aliases:
NAT15
Chromosome Location:
Chr.25: 2767265 - 2784702 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03241742_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAA60
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018040854.1 XP_017896343.1 4 - 5 531 84
GenBank mRNA JI861735.1 - 3 - 4 334 84
JO428420.1 - 4 - 5 569 84
JO622048.2 - 4 - 5 519 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0