Assay Details
Gene Symbol: TP53I11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059208.1 | XP_017914697.1 | 4 - 5 | 467 | 60 | |
XM_018059209.1 | XP_017914698.1 | 4 - 5 | 472 | 60 | ||
XM_018059210.1 | XP_017914699.1 | 1 - 2 | 42 | 60 | ||
GenBank mRNA | KA347621.1 | - | 4 - 5 | 2853 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
