This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03250163_g1
See other TRMT10B GE Assays ›
Gene Symbol:
TRMT10B
Gene Name:
tRNA methyltransferase 10B
Gene Aliases:
-
Chromosome Location:
Chr.8: 61523123 - 61535058 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03250163_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRMT10B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005683886.3 XP_005683943.2 7 - 8 843 63
XM_018052190.1 XP_017907679.1 7 - 8 763 63
GenBank mRNA JO552385.2 - 1 - 2 94 63
KA309191.1 - 7 - 8 812 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

RNA methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline