This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03254413_m1
See other PCMTD1 GE Assays ›
Gene Symbol:
PCMTD1
Gene Name:
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.14: 61001517 - 61049880 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03254413_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCMTD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005688965.3 XP_005689022.1 8 - 9 1237 59
XM_005688967.3 XP_005689024.1 7 - 8 1110 59
XM_005688969.3 XP_005689026.1 6 - 7 1086 59
XM_013969159.2 XP_013824613.1 8 - 9 1257 59
XM_013969160.2 XP_013824614.1 7 - 8 1233 59
XM_013969161.2 XP_013824615.1 6 - 7 1219 59
XM_018058517.1 XP_017914006.1 6 - 7 1186 59
XM_018058518.1 XP_017914007.1 8 - 9 1204 59
XM_018058519.1 XP_017914008.1 7 - 8 1164 59
XM_018058520.1 XP_017914009.1 7 - 8 1057 59
XM_018058521.1 XP_017914010.1 5 - 6 936 59
XM_018058522.1 XP_017914011.1 6 - 7 792 59
XM_018058523.1 XP_017914012.1 4 - 5 604 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-q25s8:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline