This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03254983_g1
See other EFEMP1 GE Assays ›
Gene Symbol:
EFEMP1
Gene Name:
EGF containing fibulin extracellular matrix protein 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 38093731 - 38165205 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03254983_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EFEMP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018055262.1 XP_017910751.1 4 - 5 653 58
XM_018055263.1 XP_017910752.1 4 - 5 713 58
XM_018055264.1 XP_017910753.1 4 - 5 835 58
XM_018055265.1 XP_017910754.1 4 - 5 666 58
XM_018055266.1 XP_017910755.1 4 - 5 653 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

extracellular matrix structural protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0