This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03255460_m1
See other EXTL3 GE Assays ›
Gene Symbol:
EXTL3
Gene Name:
exostosin like glycosyltransferase 3
Gene Aliases:
-
Chromosome Location:
Chr.8: 9832499 - 9965159 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03255460_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EXTL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018051858.1 XP_017907347.1 4 - 5 2954 61
XM_018051859.1 XP_017907348.1 5 - 6 3068 61
XM_018051860.1 XP_017907349.1 4 - 5 2961 61
GenBank mRNA JT650000.1 - 10 - 11 2173 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0