This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03260456_g1
See other PAQR6 GE Assays ›
Gene Symbol:
PAQR6
Gene Name:
progestin and adipoQ receptor family member 6
Gene Aliases:
-
Chromosome Location:
Chr.3: 105568861 - 105579350 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03260456_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PAQR6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018046285.1 XP_017901774.1 5 - 6 628 70
XM_018046286.1 XP_017901775.1 5 - 6 1560 70
XM_018046287.1 XP_017901776.1 5 - 6 628 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0