This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03261858_m1
See other TRAPPC2L GE Assays ›
Gene Symbol:
TRAPPC2L
Gene Name:
trafficking protein particle complex 2 like
Gene Aliases:
-
Chromosome Location:
Chr.18: 15434826 - 15439944 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03261858_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAPPC2L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018061864.1 XP_017917353.1 3 - 4 363 61
XM_018061865.1 XP_017917354.1 3 - 4 304 61
GenBank mRNA JO578964.1 - 3 - 4 408 61
JO643282.2 - 3 - 4 507 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0