This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03264032_g1
See other RBM17 GE Assays ›
Gene Symbol:
RBM17 LOC102174476
Gene Name:
RNA binding motif protein 17
splicing factor 45-like
Gene Aliases:
-
-
Chromosome Location:
Chr.13: 10866089 - 10888755 on Build ARS1
Chromosome Location:
Chr.10: 84457707 - 84462172 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
-
Assay ID Ch03264032_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018056960.1 XP_017912449.1 11 - 12 1257 96
GenBank mRNA JI861685.1 - 10 - 11 1103 96
JO551837.2 - 1 - 2 247 96
Gene Symbol: LOC102174476
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018053577.1 XP_017909066.1 2 1205 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline