This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03269006_g1
See other GPR137 GE Assays ›
Gene Symbol:
GPR137
Gene Name:
G protein-coupled receptor 137
Gene Aliases:
-
Chromosome Location:
Chr.29: 43010032 - 43013768 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03269006_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR137
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005699853.3 XP_005699910.1 1 - 2 1156 60
XM_005699854.3 XP_005699911.1 1 - 2 1156 60
XM_013976044.2 XP_013831498.1 1 - 2 1156 60
GenBank mRNA JO434022.2 - 2 - 3 654 60
JR132136.1 - 3 - 4 698 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0