This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03270050_g1
See other RPE GE Assays ›
Gene Symbol:
RPE
Gene Name:
ribulose-5-phosphate-3-epimerase
Gene Aliases:
-
Chromosome Location:
Chr.2: 38025648 - 38055618 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03270050_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005676461.3 XP_005676518.1 2 - 3 237 69
XM_005676462.3 XP_005676519.1 2 - 3 294 69
XM_013971562.2 XP_013827016.1 2 - 3 189 69
XM_013971565.2 XP_013827019.1 3 - 4 225 69
XM_013971570.2 XP_013827024.1 3 - 4 220 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_013971565.2, XM_013971570.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0