This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03272009_g1
See other NABP2 GE Assays ›
Gene Symbol:
NABP2
Gene Name:
nucleic acid binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.5: 56473403 - 56479392 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03272009_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NABP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005709487.3 XP_005709544.3 2 - 3 384 69
XM_018048089.1 XP_017903578.1 3 - 4 405 69
XM_018048090.1 XP_017903579.1 2 - 3 210 69
XM_018048091.1 XP_017903580.1 2 - 3 1109 69
GenBank mRNA JO612398.2 - 5 - 6 963 69
KA348067.1 - 2 - 3 381 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0