This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03277999_m1
See other LIPA GE Assays ›
Gene Symbol:
LIPA
Gene Name:
lipase A, lysosomal acid type
Gene Aliases:
-
Chromosome Location:
Chr.26: 40361720 - 40403635 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03277999_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018041263.1 XP_017896752.1 2 - 3 324 65
XM_018041264.1 XP_017896753.1 2 - 3 210 65
GenBank mRNA JO341066.2 - 8 - 9 2647 65
JO580031.1 - 2 - 3 239 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0