This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03336691_m1
See other YTHDF3 GE Assays ›
Gene Symbol:
YTHDF3
Gene Name:
YTH N6-methyladenosine RNA binding protein 3
Gene Aliases:
-
Chromosome Location:
Chr.14: 53953166 - 53987656 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03336691_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YTHDF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018058463.1 XP_017913952.1 6 - 7 2192 127
XM_018058464.1 XP_017913953.1 5 - 6 2204 127
XM_018058465.1 XP_017913954.1 4 - 5 2058 127

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7kcm7:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0