This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch03816102_m1
See other RARG GE Assays ›
Gene Symbol:
RARG
Gene Name:
retinoic acid receptor gamma
Gene Aliases:
-
Chromosome Location:
Chr.5: 26507575 - 26528192 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch03816102_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RARG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018047689.1 XP_017903178.1 8 - 9 1439 67
XM_018047690.1 XP_017903179.1 8 - 9 1449 67
XM_018047691.1 XP_017903180.1 7 - 8 1375 67
XM_018047692.1 XP_017903181.1 6 - 7 1497 67
GenBank mRNA KA349815.1 - 10 - 11 1812 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0