This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04283592_g1
See other LOC102173687 GE Assays ›
Gene Symbol:
LOC102173687 MAP3K3
Gene Name:
translational activator of cytochrome c oxidase 1
mitogen-activated protein kinase kinase kinase 3
Gene Aliases:
-
-
Chromosome Location:
Chr.19: 47517630 - 47536941 on Build ARS1
Chromosome Location:
Chr.19: 47521427 - 47589482 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
-
Assay ID Ch04283592_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102173687
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005693983.3 XP_005694040.1 1 - 2 466 68
XM_013972087.2 XP_013827541.1 1 - 2 466 68
XM_013972088.2 XP_013827542.1 1 - 2 466 68
XM_013972089.2 XP_013827543.1 1 - 2 466 68
GenBank mRNA JO398241.2 - 1 - 2 473 68
Gene Symbol: MAP3K3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA JT747640.1 - 1 - 2 189 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline