This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04293053_m1
See other CBLL1 GE Assays ›
Gene Symbol:
CBLL1
Gene Name:
Cbl proto-oncogene like 1
Gene Aliases:
-
Chromosome Location:
Chr.4: 71501624 - 71518068 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04293053_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CBLL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005679137.3 XP_005679194.1 5 - 6 784 88
XM_005679138.3 XP_005679195.1 5 - 6 783 88
GenBank mRNA JO581587.1 - 5 - 6 616 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0