This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04653713_g1
See other GPX4 GE Assays ›
Gene Symbol:
GPX4
Gene Name:
glutathione peroxidase 4
Gene Aliases:
-
Chromosome Location:
Chr.7: 66891528 - 66893833 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04653713_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPX4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285712.2 NP_001272641.2 3 - 4 410 63
GenBank mRNA GQ302986.3 - 3 - 4 319 63
GU131344.2 - 3 - 4 319 63
JO445263.2 - 4 - 5 497 63
JT762899.1 - 4 - 5 483 63
KA341887.1 - 4 - 5 482 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

peroxidase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0