This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04654523_g1
See other HMOX1 GE Assays ›
Gene Symbol:
HMOX1
Gene Name:
heme oxygenase 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 72304175 - 72311947 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04654523_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HMOX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285567.1 NP_001272496.1 4 - 5 732 74
GenBank mRNA JF731041.1 - 4 - 5 732 74
JO580392.1 - 4 - 5 902 74
KA334415.1 - 4 - 5 884 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

oxygenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0